Pyrazolo[1,5-a]pyridine-7-carbaldehyde 97%
Leverantör: Apollo Scientific
OR480774-1G OR480774-5G
APOSOR480774-1GEA 2550 SEK
APOSOR480774-1G APOSOR480774-5G
Pyrazolo[1,5-a]pyridine-7-carbaldehyde 97%
Pyrazolo[1,5-a]pyridine-7-carbaldehyde
Formel: C₈H₆N₂O
MW: 146.15 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06739029
CAS nummer: 362661-83-4