5-Chloro-1H-indole-3-carboxylic acid 95%

Leverantör: Apollo Scientific

OR480765-10G OR480765-5G
APOSOR480765-10GEA 2800 SEK
APOSOR480765-10G APOSOR480765-5G
5-Chloro-1H-indole-3-carboxylic acid 95%
5-Chloro-1H-indole-3-carboxylic acid
Formel: C₉H₆ClNO₂
MW: 195,61 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03410308
CAS nummer: 10406-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR