3-Chloro-4-nitrobenzenesulphonyl chloride 96%
Leverantör: Apollo Scientific
Synonyms: 3-Chloro-4-nitrobenzene-1-sulphonyl chloride
OR480758-1G OR480758-5G
APOSOR480758-1GEA 3200 SEK
APOSOR480758-1G APOSOR480758-5G
3-Chloro-4-nitrobenzenesulphonyl chloride 96%
3-Chloro-4-nitrobenzenesulphonyl chloride
Formel: C₆H₃Cl₂NO₄S
MW: 256,06 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04973767
CAS nummer: 64835-30-9