Ethyl-2-amino-4,5,6,7-tetrahydrobenzo[d]thiazole-6-carboxylate 95%
Leverantör: Apollo Scientific
Synonyms:
Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[d]thiazole-6-carboxylate
OR480718-10G
OR480718-5G
APOSOR480718-10GEA
8725
SEK
APOSOR480718-10G
APOSOR480718-5G
Ethyl-2-amino-4,5,6,7-tetrahydrobenzo[d]thiazole-6-carboxylate 95%
Ethyl-2-amino-4,5,6,7-tetrahydrobenzo[d]thiazole-6-carboxylate
Formel:
C₁₀H₁₄N₂O₂S MW: 226,3 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD13195375 CAS nummer: 134136-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...