2,4-Dichlorothieno[2,3-d]pyrimidine 98%
Leverantör: Apollo Scientific
OR480713-1G OR480713-25G OR480713-5G
APOSOR480713-1GEA 380 SEK
APOSOR480713-1G APOSOR480713-25G APOSOR480713-5G
2,4-Dichlorothieno[2,3-d]pyrimidine 98%
2,4-Dichlorothieno[2,3-d]pyrimidine
Formel: C₆H₂Cl₂N₂S
MW: 205,0645 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09743991
CAS nummer: 18740-39-1