1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 98%
Leverantör: Apollo Scientific
Synonyms: 1-Benzyl N-Boc-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic Acid 1-Benzyl Ester, BOC-L-Aspartic acid 1-benzyl ester, N-Boc-L-aspartic Acid 1-Benzyl Ester, Boc-Asp-OBzl, Boc-Asp-Obzl
OR480637-25G OR480637-100G
APOSOR480637-25GEA 1300 SEK
APOSOR480637-25G APOSOR480637-100G
1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 98%
1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate
Formel: C₁₆H₂₁NO₆
MW: 323,35 g/mol
Smältpunkt: 96…100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00065563
CAS nummer: 30925-18-9