o-Phenylenediacetonitrile 98%

Leverantör: Apollo Scientific

Synonyms: 1,2-Phenylenediacetonitrile, 1,2-Bis(cyanomethyl)-benzene

OR480625-100G OR480625-50G
APOSOR480625-100GEA 2950 SEK
APOSOR480625-100G APOSOR480625-50G
o-Phenylenediacetonitrile 98%
o-Phenylenediacetonitrile
Formel: C₁₀H₈N₂
MW: 156,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00001905
CAS nummer: 613-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR