N-(tert-Butoxycarbonyl)-L-tert-leucine 98%
Leverantör: Apollo Scientific
Varning
Synonyms: N-Boc-L-tert-leucine, Boc-Tle-OH, (S)-2-(Boc-amino)-3,3-dimethylbutyric Acid, (S)-2-(tert-Butoxycarbonylamino)-3,3-dimethylbutyric Acid
OR480602-25G OR480602-5G OR480602-100G
APOSOR480602-25GEA 650 SEK
APOSOR480602-25G APOSOR480602-5G APOSOR480602-100G
N-(tert-Butoxycarbonyl)-L-tert-leucine 98%
N-(tert-Butoxycarbonyl)-L-tert-leucine
Formel: C₁₁H₂₁NO₄
MW: 231.29 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00065574
CAS nummer: 62965-35-9