(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 97%
Leverantör: Apollo Scientific
Synonyms: H-D-Ser-OMe·HCl, H-D-Ser-OMe.HCl, D-Serine methyl ester hydrochloride
OR480598-100G OR480598-250G
APOSOR480598-100GEA 1425 SEK
APOSOR480598-100G APOSOR480598-250G
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 97%
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride
Formel: C₄H₉O₃N·HCl
MW: 155,6 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00066121
CAS nummer: 5874-57-7