N-Phenylglycine 97%
Leverantör: Apollo Scientific
Synonyms: 2-Anilinoacetic acid, 2-(Phenylamino)acetic acid
OR480573-100G OR480573-25G OR480573-250G OR480573-500G
APOSOR480573-100GEA 960 SEK
APOSOR480573-100G APOSOR480573-25G APOSOR480573-250G APOSOR480573-500G
N-Phenylglycine 97%
N-Phenylglycine
Formel: C₈H₉NO₂
MW: 151,17 g/mol
Smältpunkt: 121…127 °C
Storage Temperature: Omgivande
MDL Number: MFCD00014009
CAS nummer: 103-01-5