2-Amino-5-bromo-4-methoxypyridine 95%

Leverantör: Apollo Scientific

Synonyms: 5-Bromo-4-methoxypyridin-2-amine

OR47918-1G OR47918-250MG
APOSOR47918-1GEA 3450 SEK
APOSOR47918-1G APOSOR47918-250MG
2-Amino-5-bromo-4-methoxypyridine 95%
2-Amino-5-bromo-4-methoxypyridine
Formel: C₆H₇BrN₂O
MW: 203,0400 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD15143383
CAS nummer: 1232431-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR