Aminoacetaldehyde dimethyl acetal 99%
Leverantör: Apollo Scientific
Fara
Synonyms: 2,2-Dimethoxyethanamine
OR4745-100G OR4745-25G OR4745-500G
APOSOR4745-100GEA 515 SEK
APOSOR4745-100G APOSOR4745-25G APOSOR4745-500G
Aminoacetaldehyde dimethyl acetal 99%
Aminoacetaldehyde dimethyl acetal
Formel: C₄H₁₁NO₂
Kokpunkt: 135…139 °C/95mmHg
Densitet: 0,965 g/cm³ (25 °C)
Flampunkt: 44 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008135
CAS nummer: 22483-09-6
EINECS: 245-026-5
UN: 2733
ADR: 3+8,III