3-Methylbenzylamine 97%

Leverantör: Apollo Scientific
Fara

Synonyms: m-Tolylmethanamine, 3-(Aminomethyl)toluene

OR4723-5G OR4723-25G
APOSOR4723-5GEA 805 SEK
APOSOR4723-5G APOSOR4723-25G
3-Methylbenzylamine 97%
3-Methylbenzylamine
Formel: C₈H₁₁N
MW: 121,18 g/mol
Kokpunkt: 202…205 °C
Densitet: 0,966 g/cm³ (25 °C)
Flampunkt: 81 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008118
CAS nummer: 100-81-2
EINECS: 202-890-8
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR