1,3,5-Triformylbenzene 97%

Leverantör: Apollo Scientific

Synonyms: 1,3,5-benzenetricarbaldehyde, Trimesic formaldehyde, Benzene-1,3,5-tricarbaldehyde

OR471671-1G OR471671-5G
APOSOR471671-1GEA 1425 SEK
APOSOR471671-1G APOSOR471671-5G
1,3,5-Triformylbenzene 97%
1,3,5-Triformylbenzene
Formel: C₉H₆O₃
MW: 162,14 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00182470
CAS nummer: 3163-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR