4-Chlorotetrahydropyran 98%
Leverantör: Apollo Scientific
OR471653-10G
OR471653-5G
APOSOR471653-10GEA
1600
SEK
APOSOR471653-10G
APOSOR471653-5G
4-Chlorotetrahydropyran 98%
4-Chlorotetrahydropyran
Formel:
C₅H₉ClO MW: 120,5800 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00074911 CAS nummer: 1768-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...