(4-Nitrophenyl)methanesulphonyl chloride, Technical Grade
Leverantör: Apollo Scientific
Fara
Synonyms: 4-Nitro-α-toluenesulphonyl chloride
OR4716-1G OR4716-250MG OR4716-5G
APOSOR4716-1GEA 855 SEK
APOSOR4716-1G APOSOR4716-250MG APOSOR4716-5G
(4-Nitrophenyl)methanesulphonyl chloride, Technical Grade
(4-Nitrophenyl)methanesulphonyl chloride
Formel: C₇H₆ClNO₄S
MW: 235.64 g/mol
Smältpunkt: 90…93 °C
Storage Temperature: Omgivande
MDL Number: MFCD00034030
CAS nummer: 4025-75-6
UN: 3261
ADR: 8 II