2-Isobutylaniline

Leverantör: Apollo Scientific

OR471289-1G OR471289-250MG OR471289-5G
APOSOR471289-1GEA 5525 SEK
APOSOR471289-1G APOSOR471289-250MG APOSOR471289-5G
2-Isobutylaniline
2-Isobutylaniline
Formel: C₁₀H₁₅N
Storage Temperature: Omgivande
CAS nummer: 71182-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR