3',5'-Dibromoacetophenone

Leverantör: Apollo Scientific

Synonyms: 1-(3,5-Dibromophenyl)ethanone

OR470501-1G OR470501-5G
APOSOR470501-1GEA 705 SEK
APOSOR470501-1G APOSOR470501-5G
3',5'-Dibromoacetophenone
3',5'-Dibromoacetophenone
Formel: C₈H₆Br₂O
MW: 277,9400 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00078665
CAS nummer: 14401-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR