6-(2-Methoxyphenyl)nicotinic acid

Leverantör: Apollo Scientific

OR470336-1G OR470336-5G OR470336-250MG
APOSOR470336-1GEA 5825 SEK
APOSOR470336-1G APOSOR470336-5G APOSOR470336-250MG
6-(2-Methoxyphenyl)nicotinic acid
6-(2-Methoxyphenyl)nicotinic acid
Formel: C₁₃H₁₁NO₃
MW: 229.24 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06410400
CAS nummer: 887976-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR