Methyl-5-bromo-2-(methylsulphonyl)-4-pyrimidinecarboxylate

Leverantör: Apollo Scientific

OR470078-5G
APOSOR470078-5GEA 11875 SEK
APOSOR470078-5G
Methyl-5-bromo-2-(methylsulphonyl)-4-pyrimidinecarboxylate
Methyl-5-bromo-2-(methylsulphonyl)-4-pyrimidinecarboxylate
Formel: C₇H₇BrN₂O₄S
MW: 295.12 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11100780
CAS nummer: 1060795-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR