tert-Butyl-3-bromo-5,6-dihydro[1,2,4]triazolo[4,3-a]pyrazine-7(8H)-carboxylate
Leverantör: Apollo Scientific
Varning
Synonyms: tert-Butyl 3-bromo-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazine-7(8H)-carboxylate, 7-Boc-3-bromo-5,6,7,8-tetrahydro-1,2,4-triazolo[4,3-a]pyrazine
OR470061-1G OR470061-250MG OR470061-5G
APOSOR470061-1GEA 2000 SEK
APOSOR470061-1G APOSOR470061-250MG APOSOR470061-5G
tert-Butyl-3-bromo-5,6-dihydro[1,2,4]triazolo[4,3-a]pyrazine-7(8H)-carboxylate
tert-Butyl-3-bromo-5,6-dihydro[1,2,4]triazolo[4,3-a]pyrazine-7(8H)-carboxylate
Formel: C₁₀H₁₅BrN₄O₂
MW: 303,16 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08448169
CAS nummer: 723286-80-4