3,5-Dimethyl-1,2-benzenediamine
Leverantör: Apollo Scientific
OR470020-1G
OR470020-5G
APOSOR470020-1GEA
1125
SEK
APOSOR470020-1G
APOSOR470020-5G
3,5-Dimethyl-1,2-benzenediamine
3,5-Dimethyl-1,2-benzenediamine
Formel:
C₈H₁₂N₂ Storage Temperature: Omgivande |
MDL Number:
MFCD00972722 CAS nummer: 3171-46-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...