2,3-Dihydroimidazo[2,1-b][1,3]benzothiazol-6-amine
Leverantör: Apollo Scientific
OR46728-1G OR46728-250MG OR46728-500MG OR46728-5G
APOSOR46728-1GEA 925 SEK
APOSOR46728-1G APOSOR46728-250MG APOSOR46728-500MG APOSOR46728-5G
2,3-Dihydroimidazo[2,1-b][1,3]benzothiazol-6-amine
2,3-Dihydroimidazo[2,1-b][1,3]benzothiazol-6-amine
Formel: C₉H₉N₃S
MW: 191.26 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00044717
CAS nummer: 78291-99-3