2-Cyclopropylisonicotinonitrile

Leverantör: Apollo Scientific

OR46710-1G OR46710-100MG OR46710-250MG OR46710-500MG OR46710-2.5G
APOSOR46710-1GEA 6000 SEK
APOSOR46710-1G APOSOR46710-100MG APOSOR46710-250MG APOSOR46710-500MG APOSOR46710-2.5G
2-Cyclopropylisonicotinonitrile
2-Cyclopropylisonicotinonitrile
Formel: C₉H₈N₂
MW: 144.18 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18089268
CAS nummer: 1020747-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR