4-Methoxycyclohexanecarbonyl chloride
Leverantör: Apollo Scientific
OR46690-1G OR46690-250MG OR46690-500MG
APOSOR46690-1GEA 5900 SEK
APOSOR46690-1G APOSOR46690-250MG APOSOR46690-500MG
4-Methoxycyclohexanecarbonyl chloride
4-Methoxycyclohexanecarbonyl chloride
Formel: C₈H₁₃ClO₂
Storage Temperature: Omgivande
MDL Number: MFCD03424700
CAS nummer: 123790-13-6