Nicotinoyl chloride hydrochloride 95%
Leverantör: Apollo Scientific
Fara
OR46575-100G OR46575-25G
APOSOR46575-100GEA 1900 SEK
APOSOR46575-100G APOSOR46575-25G
Nicotinoyl chloride hydrochloride 95%
Nicotinoyl chloride hydrochloride
Formel: C₆H₅Cl₂NO
MW: 178,02 g/mol
Smältpunkt: 151…157 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012820
CAS nummer: 20260-53-1
UN: 3261
ADR: 8,II