(R)-(+)-Hydroxypyrrolidine
Leverantör: Apollo Scientific
Varning
Synonyms: (R)-Pyrrolidin-3-ol, (R)-(+)-3-Pyrrolidinol, (R)-(+)-3-Hydroxypyrrolidine
OR4628-1G OR4628-5G OR4628-250MG
APOSOR4628-1GEA 650 SEK
APOSOR4628-1G APOSOR4628-5G APOSOR4628-250MG
(R)-(+)-Hydroxypyrrolidine
(R)-(+)-Hydroxypyrrolidine
Chiral intermediate for pharmaceuticals and agrochemicals.
Formel: C₄H₉NO
MW: 87,12 g/mol
Kokpunkt: 215…216 °C
Smältpunkt: 15 °C
Densitet: 1,078 g/cm³ (25 °C)
Flampunkt: 113 °C
Storage Temperature: Omgivande
MDL Number: MFCD00145220
CAS nummer: 2799-21-5