(S)-(-)-Hydroxypyrrolidine 97%
Leverantör: Apollo Scientific
Varning
Synonyms: (S)-(-)-3-Pyrrolidinol, (S)-Pyrrolidin-3-ol, (S)-(-)-3-Hydroxypyrrolidine
OR4627-1G OR4627-5G OR4627-25G
APOSOR4627-1GEA 380 SEK
APOSOR4627-1G APOSOR4627-5G APOSOR4627-25G
(S)-(-)-Hydroxypyrrolidine 97%
(S)-(-)-Hydroxypyrrolidine
Chiral intermediate for pharmaceuticals and agrochemicals.
Formel: C₄H₉NO
MW: 87,12 g/mol
Kokpunkt: 108…110 °C/8mmHg
Smältpunkt: 15 °C
Densitet: 1,048 g/cm³ (25 °C)
Flampunkt: 110 °C
Storage Temperature: Frys
MDL Number: MFCD00192426
CAS nummer: 100243-39-8