(3S)-(+)-1-Benzyl-3-aminopyrrolidine
Leverantör: Apollo Scientific
Fara
Synonyms: (S)-1-Benzyl-3-aminopyrrolidine, (S)-(+)-1-Benzyl-3-aminopyrrolidine
OR4623-5G OR4623-25G
APOSOR4623-5GEA 1550 SEK
APOSOR4623-5G APOSOR4623-25G
(3S)-(+)-1-Benzyl-3-aminopyrrolidine
(3S)-(+)-1-Benzyl-3-aminopyrrolidine
Chiral intermediate for pharmaceuticals and agrochemicals.
Formel: C₁₁H₁₆N₂
MW: 176,26 g/mol
Kokpunkt: 100…105 °C/3mmHg
Densitet: 1,02 g/cm³ (25 °C)
Flampunkt: 140 °C
Storage Temperature: Omgivande
MDL Number: MFCD00082766
CAS nummer: 114715-38-7
UN: 2735
ADR: 8,III