Methyl-5-bromopyrimidine-2-carboxyate
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
Methyl 5-Bromopyrimidine-2-carboxylate, 5-Bromopyrimidine-2-carboxylic acid methyll ester
OR46221-1G
OR46221-5G
APOSOR46221-1GEA
720
SEK
APOSOR46221-1G
APOSOR46221-5G
Methyl-5-bromopyrimidine-2-carboxyate
Methyl-5-bromopyrimidine-2-carboxyate
Formel:
C₆H₅BrN₂O₂ MW: 217,02 g/mol Smältpunkt: 148…149 °C Storage Temperature: Omgivande |
MDL Number:
MFCD11111606 CAS nummer: 89581-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...