(R)-(+)-β-Hydroxy-γ-butyrolactone 90%, Technical Grade
Leverantör: Apollo Scientific
Varning
Synonyms: (R)-4-Hydroxydihydrofuran-2(3H)-one
OR4605-1G OR4605-5G OR4605-10G
APOSOR4605-1GEA 1625 SEK
APOSOR4605-1G APOSOR4605-5G APOSOR4605-10G
(R)-(+)-β-Hydroxy-γ-butyrolactone 90%, Technical Grade
(R)-(+)-β-Hydroxy-γ-butyrolactone
Formel: C₄H₆O₃
MW: 102,089 g/mol
Kokpunkt: 98…100 °C/0,3mmHg
Densitet: 1,302 g/cm³ (25 °C)
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00211248
CAS nummer: 58081-05-3