Resveratrol 99%
Leverantör: Apollo Scientific
Varning
Synonyms: trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol
OR46018-100G OR46018-500G OR46018-1KG
APOSOR46018-100GEA 1175 SEK
APOSOR46018-100G APOSOR46018-500G APOSOR46018-1KG
Resveratrol 99%
Resveratrol
Formel: C₁₄H₁₂O₃
MW: 228,25 g/mol
Smältpunkt: 261…267 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00133799
CAS nummer: 501-36-0