N-(3-Bromopropyl)phthalimide

Leverantör: Apollo Scientific
Varning

OR46012-100G OR46012-25G OR46012-5G
APOSOR46012-100GEA 1425 SEK
APOSOR46012-100G APOSOR46012-25G APOSOR46012-5G
N-(3-Bromopropyl)phthalimide
N-(3-Bromopropyl)phthalimide
Formel: C₁₁H₁₀BrNO₂
MW: 268,11 g/mol
Smältpunkt: 72…74 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005904
CAS nummer: 5460-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR