2-Jodpropan 99%

Leverantör: Apollo Scientific
Varning

Synonyms: Isopropyl iodide

OR4540-100G OR4540-500G
APOSOR4540-100GEA 1050 SEK
APOSOR4540-100G APOSOR4540-500G
2-Jodpropan 99%
2-Jodpropan
Formel: C₃H₇I
MW: 169,99 g/mol
Kokpunkt: 88…90 °C
Smältpunkt: –90 °C
Densitet: 1,703 g/cm³ (25 °C)
Flampunkt: 42 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001071
CAS nummer: 75-30-9
EINECS: 200-562-9
UN: 2392
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR