3-(Methoxymethyl)piperidine hydrochloride 97%
Leverantör: Apollo Scientific
OR452125-1G OR452125-500MG
APOSOR452125-1GEA 4625 SEK
APOSOR452125-1G APOSOR452125-500MG
3-(Methoxymethyl)piperidine hydrochloride 97%
3-(Methoxymethyl)piperidine hydrochloride
Formel: C₇H₁₆ClNO
MW: 165.66 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08166754
CAS nummer: 688809-97-4