4-Benzylpiperidine-1,4-dicarboxylic acid mono-tert-butyl ester 97%
Leverantör: Apollo Scientific
Synonyms: 4-Benzyl-1-(tert-butoxycarbonyl)piperidine-4-carboxylic acid, 1-Boc-4-benzyl-4-piperidinecarboxylic acid
OR452118-1G OR452118-250MG OR452118-500MG
APOSOR452118-1GEA 7725 SEK
APOSOR452118-1G APOSOR452118-250MG APOSOR452118-500MG
4-Benzylpiperidine-1,4-dicarboxylic acid mono-tert-butyl ester 97%
4-Benzylpiperidine-1,4-dicarboxylic acid mono-tert-butyl ester
Formel: C₁₈H₂₅NO₄
MW: 319,4 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02179131
CAS nummer: 167263-11-8