1-(Chloromethyl)-4-nitro-1H-pyrazole
Leverantör: Apollo Scientific
Varning
OR451411-1G OR451411-5G
APOSOR451411-1GEA 2325 SEK
APOSOR451411-1G APOSOR451411-5G
1-(Chloromethyl)-4-nitro-1H-pyrazole
1-(Chloromethyl)-4-nitro-1H-pyrazole
Formel: C₄H₄ClN₃O₂
Storage Temperature: Omgivande
MDL Number: MFCD03423394
CAS nummer: 51355-88-5