(R)-3-Methoxypyrrolidine hydrochloride
Leverantör: Apollo Scientific
Varning
OR451217-1G OR451217-5G
APOSOR451217-1GEA 1025 SEK
APOSOR451217-1G APOSOR451217-5G
(R)-3-Methoxypyrrolidine hydrochloride
(R)-3-Methoxypyrrolidine hydrochloride
Formel: C₅H₁₂ClNO
Storage Temperature: Omgivande
MDL Number: MFCD11849816
CAS nummer: 474707-30-7