(R)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
Leverantör: Apollo Scientific
Synonyms: (R)-(1,2,3,4-Tetrahydroisoquinolin-3-yl)methanol
OR451146-1G OR451146-25G OR451146-5G
APOSOR451146-1GEA 1725 SEK
APOSOR451146-1G APOSOR451146-25G APOSOR451146-5G
(R)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
(R)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
Formel: C₁₀H₁₃NO
MW: 163.22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02684327
CAS nummer: 62855-02-1