(Bromomethyl)cyclobutane 98%
Leverantör: Apollo Scientific
Varning
Synonyms: Cyclobutyl methyl bromide
OR451132-1G OR451132-5G OR451132-25G
APOSOR451132-1GEA 245 SEK
APOSOR451132-1G APOSOR451132-5G APOSOR451132-25G
(Bromomethyl)cyclobutane 98%
(Bromomethyl)cyclobutane
Formel: C₅H₉Br
MW: 149,03 g/mol
Kokpunkt: 145…148 °C
Densitet: 1,339 g/cm³ (25 °C)
Flampunkt: 27 °C
Storage Temperature: Omgivande
MDL Number: MFCD00040960
CAS nummer: 17247-58-4
UN: 1993
ADR: 3,III