4-(2-Bromoethyl)tetrahydropyran
Leverantör: Apollo Scientific
|
Fara
|
OR451047-1G
OR451047-5G
APOSOR451047-1GEA
1275
SEK
APOSOR451047-1G
APOSOR451047-5G
4-(2-Bromoethyl)tetrahydropyran
4-(2-Bromoethyl)tetrahydropyran
Formel:
C₇H₁₃BrO MW: 193,0800 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD09800414 CAS nummer: 4677-20-7 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...