(±)-Boc-α-phosphonoglycine trimethyl ester 98%

Leverantör: Apollo Scientific
Fara

Synonyms: Methyl 2-((tert-butoxycarbonyl)amino)-2-(dimethoxyphosphoryl)acetate

OR45088-25G OR45088-1G OR45088-5G
APOSOR45088-25GEA 1325 SEK
APOSOR45088-25G APOSOR45088-1G APOSOR45088-5G
(±)-Boc-α-phosphonoglycine trimethyl ester 98%
(±)-Boc-α-phosphonoglycine trimethyl ester
Formel: C₁₀H₂₀NO₇P
MW: 297,24 g/mol
Smältpunkt: 66…69 °C
Storage Temperature: Omgivande
MDL Number: MFCD02178893
CAS nummer: 89524-98-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR