Ethyl-4-bromo-1,5-dimethyl-1H-pyrazole-3-carboxylate
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
Ethyl 4-Bromo-1,5-dimethyl-1H-pyrazole-3-carboxylate
OR45002-1G
OR45002-5G
APOSOR45002-1GEA
1675
SEK
APOSOR45002-1G
APOSOR45002-5G
Ethyl-4-bromo-1,5-dimethyl-1H-pyrazole-3-carboxylate
Ethyl-4-bromo-1,5-dimethyl-1H-pyrazole-3-carboxylate
Formel:
C₈H₁₁BrN₂O₂ MW: 247,0891 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00207970 CAS nummer: 5775-90-6 |