m-Tolylacetic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-(m-Tolyl)acetic acid, (3-Methylphenyl)acetic acid
OR4364-10G OR4364-50G OR4364-100MG
APOSOR4364-10GEA 1150 SEK
APOSOR4364-10G APOSOR4364-50G APOSOR4364-100MG
m-Tolylacetic acid 98%
m-Tolylacetic acid
Formel: (C₆H₄(CH₃))CH₂COOH
MW: 150,18 g/mol
Smältpunkt: 62…66 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004340
CAS nummer: 621-36-3
EINECS: 210-683-9