5-Bromosalicylic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Bromo-2-hydroxybenzoic acid
OR43524-100G OR43524-500G OR43524-1KG
APOSOR43524-100GEA 1675 SEK
APOSOR43524-100G APOSOR43524-500G APOSOR43524-1KG
5-Bromosalicylic acid 98%
5-Bromosalicylic acid
Formel: C₇H₅BrO₃
MW: 217,02 g/mol
Smältpunkt: 162…166 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002455
CAS nummer: 89-55-4