N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester
Leverantör: Apollo Scientific
Varning
Synonyms: N-Boc-trans-4-hydroxy-L-proline Methyl Ester, (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic acid 1-tert-Butyl 2-Methyl Ester, 1-tert-Butyl 2-Methyl (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylate, Boc-Hyp-OMe, 1-(tert-Butyl) 2-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
OR43520-100G OR43520-25G OR43520-500G
APOSOR43520-100GEA 840 SEK
APOSOR43520-100G APOSOR43520-25G APOSOR43520-500G
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester
Formel: C₁₁H₁₉NO₅
MW: 245,28 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00076981
CAS nummer: 74844-91-0