2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester 96%
Leverantör: Apollo Scientific
Varning
Synonyms: 5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine-2-carboxylic acid methylamide, N-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinamide
OR4324-1G OR4324-5G
APOSOR4324-1GEA 2400 SEK
APOSOR4324-1G APOSOR4324-5G
2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester 96%
2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester
Formel: C₁₃H₁₉BN₂O₃
MW: 262.11 g/mol
Smältpunkt: 113…115 °C
Storage Temperature: Kylskåp
MDL Number: MFCD08458483
CAS nummer: 945863-21-8