3-Bromo-5-methylaniline 97%
Leverantör: Apollo Scientific
Varning
OR42210-100G OR42210-25G OR42210-5G
APOSOR42210-100GEA 10175 SEK
APOSOR42210-100G APOSOR42210-25G APOSOR42210-5G
3-Bromo-5-methylaniline 97%
3-Bromo-5-methylaniline
Formel: C7H8BrN
MW: 186,05 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD11845553
CAS nummer: 74586-53-1