2-Pyrimidinemethanol 98%

Leverantör: Apollo Scientific
Varning

Synonyms: 2-(Hydroxymethyl)pyrimidine

OR42130-25G OR42130-5G
APOSOR42130-25GEA 2650 SEK
APOSOR42130-25G APOSOR42130-5G
2-Pyrimidinemethanol 98%
2-Pyrimidinemethanol
Formel: C₅H₆N₂O
MW: 110,12 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06796201
CAS nummer: 42839-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR