10-Phenyl-9-anthraceneboronic acid 98%

Leverantör: Apollo Scientific
Varning

Synonyms: (10-Phenylanthracen-9-yl)boronic acid

OR42025-5G OR42025-10G
APOSOR42025-5GEA 1725 SEK
APOSOR42025-5G APOSOR42025-10G
10-Phenyl-9-anthraceneboronic acid 98%
10-Phenyl-9-anthraceneboronic acid
Formel: C₂₀H₁₅BO₂
MW: 298,15 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD11111989
CAS nummer: 334658-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR